JWPH942F374B J & W Pharmlab LLC

7-Methoxy-chroman-3-carboxylic acid methyl ester

  • MDL number MFCD04114632
  • Cas Number : 885271-74-9
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/33R0075

Safety Information

MSDS https://www.jwpharmlab.com/home/product/33R0075
Availability details for
Quantity
  • Availability
  • Add to Cart