JWPH942F374F J & W Pharmlab LLC

5-Chloromethyl-2-methylsulfanyl-1H-imidazole

  • MDL number MFCD10696302
  • Cas Number : 944905-27-5
  • Molecular Weight : 162.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/40R0112

Safety Information

MSDS https://www.jwpharmlab.com/home/product/40R0112
Availability details for
Quantity
  • Availability
  • Add to Cart