JWPH942F3756 J & W Pharmlab LLC

5-Methyl-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine-2-carboxylic acid methyl ester

  • MDL number MFCD18723839
  • Cas Number : 1842396-18-2
  • Molecular Weight : 277.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/32R0195P

Safety Information

MSDS https://www.jwpharmlab.com/home/product/32R0195P
Availability details for
Quantity
  • Availability
  • Add to Cart