JWPH942F378C J & W Pharmlab LLC

6,8-Dibromo-2H-chromene-3-carboxylic acid

  • MDL number MFCD04114599
  • Cas Number : 855286-71-4
  • Molecular Weight : 333.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/33R0027

Safety Information

MSDS https://www.jwpharmlab.com/home/product/33R0027
Availability details for
Quantity
  • Availability
  • Add to Cart