JWPH942F3794 J & W Pharmlab LLC

7-Methoxy-2H-chromene-3-carboxylic acid methyl ester

  • MDL number MFCD04114607
  • Cas Number : 86236-08-0
  • Molecular Weight : 220.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/33R0048

Safety Information

MSDS https://www.jwpharmlab.com/home/product/33R0048
Availability details for
Quantity
  • Availability
  • Add to Cart