JWPH942F3797 J & W Pharmlab LLC

2-(4-Fluoro-2-trifluoromethyl-phenyl)-1H-imidazole

  • MDL number MFCD08668769
  • Cas Number : 1187932-73-5
  • Molecular Weight : 230.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/40R0084

Safety Information

MSDS https://www.jwpharmlab.com/home/product/40R0084
Availability details for
Quantity
  • Availability
  • Add to Cart