JWPH942F37A1 J & W Pharmlab LLC

5-(4-Fluoro-phenyl)-4-pyridin-2-yl-1H-imidazole-2-thiol

  • MDL number MFCD04114649
  • Cas Number : 885271-83-0
  • Molecular Weight : 271.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/40R0008

Safety Information

MSDS https://www.jwpharmlab.com/home/product/40R0008
Availability details for
Quantity
  • Availability
  • Add to Cart