JWPH942F380A J & W Pharmlab LLC

(6-tert-Butoxycarbonylamino-1H-indazol-3-yl)-acetic acid

  • MDL number MFCD09835470
  • Cas Number : 1220040-13-0
  • Molecular Weight : 291.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/45R0443

Safety Information

MSDS https://www.jwpharmlab.com/home/product/45R0443
Availability details for
Quantity
  • Availability
  • Add to Cart