JWPH942F3835 J & W Pharmlab LLC

(1H-Indazol-7-yl)-hydrazine

  • MDL number MFCD22201111
  • Cas Number : 1373223-38-1
  • Molecular Weight : 148.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/45R0536

Safety Information

MSDS https://www.jwpharmlab.com/home/product/45R0536
Availability details for
Quantity
  • Availability
  • Add to Cart