JWPH942F3854 J & W Pharmlab LLC

5-Difluoromethoxy-1H-indazole-3-carboxylic acid

  • MDL number MFCD11042710
  • Cas Number : 858661-78-6
  • Molecular Weight : 228.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/45R0618

Safety Information

MSDS https://www.jwpharmlab.com/home/product/45R0618
Availability details for
Quantity
  • Availability
  • Add to Cart