JWPH942F387C J & W Pharmlab LLC

6-Chloro-3-oxo-2,3-dihydro-1H-isoindole-5-sulfonic acid amide

  • MDL number MFCD09701279
  • Cas Number : 91862-45-2
  • Molecular Weight : 246.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/48R0296

Safety Information

MSDS https://www.jwpharmlab.com/home/product/48R0296
Availability details for
Quantity
  • Availability
  • Add to Cart