JWPH942F389E J & W Pharmlab LLC

(S)-1-Boc-3-(2-Allyloxycarbonyl-2-amino-ethyl)-indole

  • MDL number MFCD06796146
  • Cas Number : 1187931-78-7
  • Molecular Weight : 344.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/48R0232

Safety Information

MSDS https://www.jwpharmlab.com/home/product/48R0232
Availability details for
Quantity
  • Availability
  • Add to Cart