JWPH942F38C1 J & W Pharmlab LLC

2-Cbz-6-chloro-3,4-dihydro-1H-isoquinoline-1-carboxylic acid

  • MDL number MFCD04114855
  • Cas Number : 1260638-63-8
  • Molecular Weight : 345.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/50R0047

Safety Information

MSDS https://www.jwpharmlab.com/home/product/50R0047
Availability details for
Quantity
  • Availability
  • Add to Cart