JWPH942F38D7 J & W Pharmlab LLC

3-(5-Fluoro-1H-indol-3-yl)-2-methyl-propylamine

  • MDL number MFCD15474912
  • Cas Number : 1220039-79-1
  • Molecular Weight : 206.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/48R0579

Safety Information

MSDS https://www.jwpharmlab.com/home/product/48R0579
Availability details for
Quantity
  • Availability
  • Add to Cart