JWPH942F38DA J & W Pharmlab LLC

(S)-3-(5-Fluoro-1H-indol-3-yl)-2-methyl-propylamine

  • MDL number MFCD15474916
  • Cas Number : 1103518-35-9
  • Molecular Weight : 206.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/48R0583

Safety Information

MSDS https://www.jwpharmlab.com/home/product/48R0583
Availability details for
Quantity
  • Availability
  • Add to Cart