JWPH942F3922 J & W Pharmlab LLC

1-(3,4-Dimethoxyphenyl)-1-tosylmethyl isocyanide

  • MDL number MFCD05864798
  • Cas Number : 943614-50-4
  • Molecular Weight : 331.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/49R0044

Safety Information

MSDS https://www.jwpharmlab.com/home/product/49R0044
Availability details for
Quantity
  • Availability
  • Add to Cart