JWPH942F3928 J & W Pharmlab LLC

2-Methyl-3-oxo-2,3-dihydro-1H-isoindole-1-carboxylic acid

  • MDL number MFCD26938383
  • Cas Number : 1350468-65-3
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/48R0829

Safety Information

MSDS https://www.jwpharmlab.com/home/product/48R0829
Availability details for
Quantity
  • Availability
  • Add to Cart