JWPH942F393D J & W Pharmlab LLC

(4-Bromo-isoquinolin-1-yl)-hydrazine

  • MDL number MFCD10000040
  • Cas Number : 27187-06-0
  • Molecular Weight : 238.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/50R0204

Safety Information

MSDS https://www.jwpharmlab.com/home/product/50R0204
Availability details for
Quantity
  • Availability
  • Add to Cart