JWPH942F3956 J & W Pharmlab LLC

(S)-3-Aminomethyl-2-Fmoc-1,2,3,4-tetrahydro-isoquinoline

  • MDL number MFCD09997810
  • Cas Number : 1187933-38-5
  • Molecular Weight : 384.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/50R0082

Safety Information

MSDS https://www.jwpharmlab.com/home/product/50R0082
Availability details for
Quantity
  • Availability
  • Add to Cart