JWPH942F3960 J & W Pharmlab LLC

5-Methyl-3,4-dihydro-1H-isoquinoline-2,6-dicarboxylic acid 2-tert-butyl ester

  • MDL number MFCD20924900
  • Cas Number : 1824291-40-8
  • Molecular Weight : 291.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/50R0398

Safety Information

MSDS https://www.jwpharmlab.com/home/product/50R0398
Availability details for
Quantity
  • Availability
  • Add to Cart