JWPH942F397E J & W Pharmlab LLC

tert-Butyl ester 8-amino-1,3,4,5-tetrahydro-benzo[c]azepine-2-carboxylate

  • MDL number MFCD12408189
  • Cas Number : 939043-41-1
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/50R0389

Safety Information

MSDS https://www.jwpharmlab.com/home/product/50R0389
Availability details for
Quantity
  • Availability
  • Add to Cart