JWPH942F399F J & W Pharmlab LLC

2-(3-Bromo-phenyl)-oxazole-4-carbaldehyde

  • MDL number MFCD06738602
  • Cas Number : 885273-03-0
  • Molecular Weight : 252.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/56R0084

Safety Information

MSDS https://www.jwpharmlab.com/home/product/56R0084
Availability details for
Quantity
  • Availability
  • Add to Cart