JWPH942F39AA J & W Pharmlab LLC

6,8-Dimethoxy-1,2,3,4-tetrahydro-isoquinoline

  • MDL number MFCD11203699
  • Cas Number : 88207-92-5
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/50R0419

Safety Information

MSDS https://www.jwpharmlab.com/home/product/50R0419
Availability details for
Quantity
  • Availability
  • Add to Cart