JWPH942F39CB J & W Pharmlab LLC

Morpholin-3-yl-acetic acid benzyl ester

  • MDL number MFCD04114887
  • Cas Number : 885273-91-6
  • Molecular Weight : 235.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/53R0003

Safety Information

MSDS https://www.jwpharmlab.com/home/product/53R0003
Availability details for
Quantity
  • Availability
  • Add to Cart