JWPH942F39D2 J & W Pharmlab LLC

3-Morpholin-3-yl-propionic acid ethyl ester

  • MDL number MFCD04114897
  • Cas Number : 885274-01-1
  • Molecular Weight : 187.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/53R0016

Safety Information

MSDS https://www.jwpharmlab.com/home/product/53R0016
Availability details for
Quantity
  • Availability
  • Add to Cart