JWPH942F39DA J & W Pharmlab LLC

4-Benzyl-2-pyrrolidin-1-ylmethyl-morpholine

  • MDL number MFCD06658347
  • Cas Number : 635699-06-8
  • Molecular Weight : 260.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/53R0052

Safety Information

MSDS https://www.jwpharmlab.com/home/product/53R0052
Availability details for
Quantity
  • Availability
  • Add to Cart