JWPH942F39DC J & W Pharmlab LLC

(R)-[1,4]Oxazepan-6-ol

  • MDL number MFCD19227234
  • Cas Number : 1022915-33-8
  • Molecular Weight : 117.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/53R0171

Safety Information

MSDS https://www.jwpharmlab.com/home/product/53R0171
Availability details for
Quantity
  • Availability
  • Add to Cart