JWPH942F39E1 J & W Pharmlab LLC

(2S)-(+)-(5,5-Dimethyl-morpholin-2-yl)-acetic acid

  • MDL number MFCD05662362
  • Cas Number : 160415-07-6
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/53R0102

Safety Information

MSDS https://www.jwpharmlab.com/home/product/53R0102
Availability details for
Quantity
  • Availability
  • Add to Cart