JWPH942F39E4 J & W Pharmlab LLC

(R)-6,6-Dimethyl-morpholine-3,4-dicarboxylic acid 4-benzyl ester

  • MDL number MFCD11053576
  • Cas Number : 1263077-96-8
  • Molecular Weight : 293.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/53R0144

Safety Information

MSDS https://www.jwpharmlab.com/home/product/53R0144
Availability details for
Quantity
  • Availability
  • Add to Cart