JWPH942F39E5 J & W Pharmlab LLC

6,6-Dimethyl-morpholine-3-carboxylic acid methyl ester

  • MDL number MFCD11053667
  • Cas Number : 1255098-56-6
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/53R0147

Safety Information

MSDS https://www.jwpharmlab.com/home/product/53R0147
Availability details for
Quantity
  • Availability
  • Add to Cart