JWPH942F39F1 J & W Pharmlab LLC

(2S)-Morpholin-2-ylmethyl-carbamic acid benzyl ester

  • MDL number MFCD20922884
  • Cas Number : 1174913-73-5
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/53R0198

Safety Information

MSDS https://www.jwpharmlab.com/home/product/53R0198
Availability details for
Quantity
  • Availability
  • Add to Cart