JWPH942F3A14 J & W Pharmlab LLC

[2-(3-Methoxy-phenyl)-oxazol-4-yl]-methanol

  • MDL number MFCD06738572
  • Cas Number : 885272-69-5
  • Molecular Weight : 205.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/56R0044

Safety Information

MSDS https://www.jwpharmlab.com/home/product/56R0044
Availability details for
Quantity
  • Availability
  • Add to Cart