JWPH942F3A1F J & W Pharmlab LLC

2-(4-Methoxy-phenyl)-oxazole-4-carboxylic acid ethyl ester

  • MDL number MFCD06738550
  • Cas Number : 78979-61-0
  • Molecular Weight : 247.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/56R0061

Safety Information

MSDS https://www.jwpharmlab.com/home/product/56R0061
Availability details for
Quantity
  • Availability
  • Add to Cart