JWPH942F3A5C J & W Pharmlab LLC

(3-Bromo-4-hydroxymethyl-isoxazol-5-yl)-methanol

  • MDL number MFCD12913763
  • Cas Number : 153954-80-4
  • Molecular Weight : 208.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/56R0364

Safety Information

MSDS https://www.jwpharmlab.com/home/product/56R0364
Availability details for
Quantity
  • Availability
  • Add to Cart