JWPH942F3A63 J & W Pharmlab LLC

5,6,7,8-Tetrahydro-4H-cyclohepta[d]isoxazol-3-ol

  • MDL number MFCD22689286
  • Cas Number : 935-91-1
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/56R0523

Safety Information

MSDS https://www.jwpharmlab.com/home/product/56R0523
Availability details for
Quantity
  • Availability
  • Add to Cart