JWPH942F3A73 J & W Pharmlab LLC

6,7-Dihydro-4H-pyrano[3,4-d]oxazole-2-carboxylic acid

  • MDL number MFCD18250694
  • Cas Number : 1260670-92-5
  • Molecular Weight : 169.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/56R0547

Safety Information

MSDS https://www.jwpharmlab.com/home/product/56R0547
Availability details for
Quantity
  • Availability
  • Add to Cart