JWPH942F3A7D J & W Pharmlab LLC

2-(4-Fluoro-phenoxymethyl)-oxirane

  • MDL number MFCD00098471
  • Cas Number : 18123-82-5
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/57R0005

Safety Information

MSDS https://www.jwpharmlab.com/home/product/57R0005
Availability details for
Quantity
  • Availability
  • Add to Cart