JWPH942F3AB1 J & W Pharmlab LLC

N-Allyloxycarbonyl-piperid-3-one

  • MDL number MFCD04115029
  • Cas Number : 885274-83-9
  • Molecular Weight : 183.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R0157

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R0157
Availability details for
Quantity
  • Availability
  • Add to Cart