JWPH942F3AD1 J & W Pharmlab LLC

5-Fluoro-2-piperidin-4-yl-1H-benzoimidazole

  • MDL number MFCD06739002
  • Cas Number : 295790-49-7
  • Molecular Weight : 219.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R0273

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R0273
Availability details for
Quantity
  • Availability
  • Add to Cart