JWPH942F3AD7 J & W Pharmlab LLC

4-(2-Methyl-4-nitro-phenoxy)-piperidine

  • MDL number MFCD08752607
  • Cas Number : 682326-39-2
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R0290

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R0290
Availability details for
Quantity
  • Availability
  • Add to Cart