JWPH942F3AF1 J & W Pharmlab LLC

4-(3-Oxo-4-phenyl-butyryl)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD11858456
  • Cas Number : 301219-11-4
  • Molecular Weight : 345.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R0470

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R0470
Availability details for
Quantity
  • Availability
  • Add to Cart