JWPH942F3B0E J & W Pharmlab LLC

Cyclopentyl-(2,2,6,6-tetramethyl-piperidin-4-yl)-amine

  • MDL number MFCD11161644
  • Cas Number : 124172-54-9
  • Molecular Weight : 224.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R0715

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R0715
Availability details for
Quantity
  • Availability
  • Add to Cart