JWPH942F3B14 J & W Pharmlab LLC

1-N-Cbz-piperidine-4-carboxylic acid hydrazide

  • MDL number MFCD06795951
  • Cas Number : 161609-80-9
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R0643

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R0643
Availability details for
Quantity
  • Availability
  • Add to Cart