JWPH942F3B3B J & W Pharmlab LLC

(6-Hydroxymethyl-piperidin-3-yl)-carbamic acid tert-butyl ester

  • MDL number MFCD12031278
  • Cas Number : 1150618-40-8
  • Molecular Weight : 230.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R1063

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R1063
Availability details for
Quantity
  • Availability
  • Add to Cart