JWPH942F3B41 J & W Pharmlab LLC

1-(1-Methyl-piperidin-3-yl)-ethanone

  • MDL number MFCD11637836
  • Cas Number : 91324-25-3
  • Molecular Weight : 141.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R0840

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R0840
Availability details for
Quantity
  • Availability
  • Add to Cart