JWPH942F3B82 J & W Pharmlab LLC

2-Ethyl-piperidin-4-one

  • MDL number MFCD09953289
  • Cas Number : 1245698-19-4
  • Molecular Weight : 127.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R1033

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R1033
Availability details for
Quantity
  • Availability
  • Add to Cart