JWPH942F3BAF J & W Pharmlab LLC

(2S,6R)-6-Phenyl-piperidine-2-carboxylic acid hydrochloride

  • MDL number MFCD18071773
  • Cas Number : 221690-70-6
  • Molecular Weight : 241.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R1219S

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R1219S
Availability details for
Quantity
  • Availability
  • Add to Cart