JWPH942F3BBD J & W Pharmlab LLC

(1R,4R)-2-Boc-5-oxo-2-aza-bicyclo[2.2.1]heptane

  • MDL number MFCD22209795
  • Cas Number : 1400808-00-5
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R1550

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R1550
Availability details for
Quantity
  • Availability
  • Add to Cart