JWPH942F3BC2 J & W Pharmlab LLC

1-Boc-4-hydroxy-4-methylaminomethyl-piperidine

  • MDL number MFCD11920519
  • Cas Number : 1073559-55-3
  • Molecular Weight : 244.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R1123

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R1123
Availability details for
Quantity
  • Availability
  • Add to Cart