JWPH942F3BD4 J & W Pharmlab LLC

6-Benzyl-6-aza-bicyclo[3.1.1]heptan-3-one

  • MDL number MFCD30181932
  • Cas Number : 1965310-39-7
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/60R1146

Safety Information

MSDS https://www.jwpharmlab.com/home/product/60R1146
Availability details for
Quantity
  • Availability
  • Add to Cart